Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035415.hg.1 |
TC01002315.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035415.hg.1 |
ENST00000264198.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035415.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197540.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197540.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197540.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197540.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197540.hg.1 |
TC0100013184.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
ENST00000264198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013184.hg.1 |
NM_024544.2 |
Refseq |