Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HFE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_202004.3 | NP_973733.1 | 2 - 3 | 273 | 112 | |
GenBank mRNA | AK092692.1 | - | 2 - 3 | 157 | 112 | |
BC017926.1 | - | 2 - 3 | 186 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |