Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022540.hg.1 |
TC09001749.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022540.hg.1 |
ENST00000265662.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022540.hg.1 |
ENST00000341511.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022540.hg.1 |
ENST00000355090.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022540.hg.1 |
ENST00000371605.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022540.hg.1 |
ENST00000398210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022540.hg.1 |
NM_001606.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022540.hg.1 |
NM_212533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000265662.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000341511.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000371605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000431584.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000459850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000479446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000487109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000614293.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
NM_001606.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
NM_212533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000265662.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000341511.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000371605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000431584.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000459850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000479446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000487109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
ENST00000614293.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
NM_001606.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
NM_212533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182612.hg.1 |
TC0900012025.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000265662.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000341511.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000371605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000431584.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000459850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000479446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000487109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000614293.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
NM_001606.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
NM_212533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000265662.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000341511.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000371605.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000431584.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000459850.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000479446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000487109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
ENST00000614293.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
NM_001606.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
NM_212533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182611.hg.1 |
TC0900012025.hg.1 |
Clariom D TC |