Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024354.hg.1 |
TC04001769.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024354.hg.1 |
ENST00000264595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024354.hg.1 |
ENST00000506853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024354.hg.1 |
NM_000027.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024354.hg.1 |
NM_001171988.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024354.hg.1 |
NR_033655.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185677.hg.1 |
ENST00000264595.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185677.hg.1 |
ENST00000502310.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185677.hg.1 |
ENST00000506853.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185677.hg.1 |
NM_000027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185677.hg.1 |
NM_001171988.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185677.hg.1 |
NR_033655.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185677.hg.1 |
ENST00000264595.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185677.hg.1 |
ENST00000502310.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185677.hg.1 |
ENST00000506853.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185677.hg.1 |
NM_000027.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185677.hg.1 |
NM_001171988.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185677.hg.1 |
NR_033655.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185677.hg.1 |
TC0400012496.hg.1 |
Clariom D TC |