Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_001303118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_001303118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_001303118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_001303118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_001303118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_001303118.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_001303119.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_001303119.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_001303119.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_001303119.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_001303119.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012973.hg.1 |
ENST00000378681.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012973.hg.1 |
ENST00000463405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012973.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012973.hg.1 |
TC10001047.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000167068.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167068.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167068.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167068.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167068.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000167068.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000167068.hg.1 |
TC1000009839.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
ENST00000378681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
ENST00000463405.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_001303118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_001303118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_001303118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_001303118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_001303118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_001303118.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_001303119.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_001303119.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_001303119.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_001303119.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_001303119.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009839.hg.1 |
NM_145314.2 |
Refseq |