Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004737.hg.1 |
ENST00000261681.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004737.hg.1 |
ENST00000555925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004737.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004737.hg.1 |
TC14000403.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000554208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000554208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000554208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000554911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000554911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000554911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000554911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000556345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000556345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000556345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000556345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000556345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000556345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000557237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000557237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000557783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000557783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
ENST00000557783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000554208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000554208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000554208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000554911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000554911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000554911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000554911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000556345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000556345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000556345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000556345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000556345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000556345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000557237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000557237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000557783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000557783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
ENST00000557783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007491.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152482.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152482.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152482.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152482.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152482.hg.1 |
ENST00000261681.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152482.hg.1 |
ENST00000555925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400152482.hg.1 |
NM_001256550.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152482.hg.1 |
NM_022474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400152482.hg.1 |
TC1400007491.hg.1 |
Clariom D TC |