Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001959.hg.1 |
ENST00000264079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001959.hg.1 |
ENST00000394321.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001959.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001959.hg.1 |
TC19000128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000594692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000594692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000594692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000594692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000596008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000596008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000596008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000596008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000596008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000596390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000596390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000596390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000596390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000596390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000598406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000598406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000598406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000598406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000598406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199062.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199062.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199062.hg.1 |
ENST00000596008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199062.hg.1 |
ENST00000598406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199062.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199062.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900199062.hg.1 |
TC1900011653.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900199062.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199062.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199062.hg.1 |
ENST00000596008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199062.hg.1 |
ENST00000598406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199062.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900199062.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000264079.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000394321.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000594692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000594692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000594692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000594692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000595860.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000596008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000596008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000596008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000596008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000596008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000596390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000596390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000596390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000596390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000596390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000598406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000598406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000598406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000598406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000598406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
ENST00000601003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011653.hg.1 |
NM_020533.2 |
Refseq |