Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000593569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000593569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000593569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000593569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000593569.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000597030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000597030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000597030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000597030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000597030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000597232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000597232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000597232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000597232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000597232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000597232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000598657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000598657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000598657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000598657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000601287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000601287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000601287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000601287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
ENST00000601287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023804.hg.1 |
ENST00000376916.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023804.hg.1 |
ENST00000389208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023804.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023804.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023804.hg.1 |
TC19001749.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
ENST00000593569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
ENST00000598657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
TC1900011208.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
ENST00000593569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
ENST00000598657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194129.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000376916.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000389208.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000593569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000593569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000593569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000593569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000593569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000597030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000597030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000597030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000597030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000597030.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000597232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000597232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000597232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000597232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000597232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000597232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000598657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000598657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000598657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000598657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000601207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000601287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000601287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000601287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000601287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
ENST00000601287.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001024656.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011208.hg.1 |
NM_001114598.1 |
Refseq |