Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_021640_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7955783 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7955779 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7955784 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7955786 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7955781 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7955778 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7955782 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7955785 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7955780 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7955777 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
218220_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11721676_PM_a_at |
- |
- |
Affymetrix Human Genome U219 Array |
11721676_a_at |
- |
- |
Affymetrix Human Genome U219 Array |
11761104_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11761104_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005660.hg.1 |
ENST00000267103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005660.hg.1 |
ENST00000545214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005660.hg.1 |
ENST00000548632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005660.hg.1 |
ENST00000548845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005660.hg.1 |
ENST00000549488.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005660.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005660.hg.1 |
TC12000436.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009266.hg.1 |
ENST00000545095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155213.hg.1 |
TC1200007710.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000550199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
ENST00000551670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007710.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000267103.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000547490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000547864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000548632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000548845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000549177.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
ENST00000549488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
NM_021640.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155214.hg.1 |
TC1200007710.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009266.hg.1 |
ENST00000545095.1 |
Ensembl |
Human Genome U133 Plus 2.0 Array |
218220_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
218220_at |
- |
- |
Human Genome U133A 2.0 Array |
218220_at |
- |
- |
Human Genome U133A 2.0 Array |
218220_PM_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_021640_at |
- |
- |
AffymetrixPathwork Diagnostics Pathchip |
pw567213 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
218220_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
218220_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
218220_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
218220_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
218220_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
218220_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
218220_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
218220_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
218220_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
218220_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
218220_at5 |
- |
- |
Human Genome U95 Set |
53911_PM_at |
- |
- |
Human Genome U95 Set |
53911_at |
- |
- |
Human Genome U95 Set |
85648_PM_at |
- |
- |
Human Genome U95 Set |
85648_at |
- |
- |
Human Genome U133 Set |
218220_PM_at |
- |
- |
Human Genome U133 Set |
218220_at |
- |
- |