Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004591.hg.1 |
TC07000306.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004591.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004591.hg.1 |
ENST00000420324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004591.hg.1 |
ENST00000430738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004591.hg.1 |
ENST00000435376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004591.hg.1 |
ENST00000539619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004591.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004592.hg.1 |
TC07000306.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004592.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004592.hg.1 |
ENST00000539619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004592.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000461741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000461741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000461741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000461741.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004593.hg.1 |
TC07000306.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004593.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004593.hg.1 |
ENST00000420324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004593.hg.1 |
ENST00000430738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004593.hg.1 |
ENST00000539619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004593.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
ENST00000461741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
ENST00000461741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151965.hg.1 |
TC0700007475.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700151966.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151966.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151966.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700151966.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700151966.hg.1 |
TC0700007475.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000348904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000420324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000430738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000435376.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000461741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000461741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000461741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000461741.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
ENST00000539619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001100159.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267865.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007475.hg.1 |
NM_001267866.1 |
Refseq |