Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012408.hg.1 |
TC06004100.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012408.hg.1 |
ENST00000237275.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012408.hg.1 |
ENST00000539295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012408.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012409.hg.1 |
TC06004100.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012409.hg.1 |
ENST00000237275.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012409.hg.1 |
ENST00000539295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012409.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199619.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199619.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199619.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199619.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199619.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600199619.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600199619.hg.1 |
TC0600014190.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000237275.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
ENST00000539295.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014190.hg.1 |
NM_001013623.2 |
Refseq |