Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016452.hg.1 |
TC09001319.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016452.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016452.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010737.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010737.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010737.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010737.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010737.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010737.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010737.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010737.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010737.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010737.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010737.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010737.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016453.hg.1 |
TC09001319.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016453.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016453.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016454.hg.1 |
TC09001319.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016454.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016454.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016451.hg.1 |
TC09001319.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016451.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016451.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173107.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173107.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173107.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173107.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173107.hg.1 |
TC0900010737.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010737.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010737.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010737.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010737.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010737.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010737.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010737.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010737.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010737.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010737.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010737.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010737.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173108.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173108.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173108.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173108.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173108.hg.1 |
TC0900010737.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173105.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173105.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173105.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173105.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173105.hg.1 |
TC0900010737.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173106.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173106.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173106.hg.1 |
ENST00000375765.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173106.hg.1 |
NM_017594.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173106.hg.1 |
TC0900010737.hg.1 |
Clariom D TC |