Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146706.1 | NP_001140178.1 | 4 - 5 | 635 | 64 | |
GenBank mRNA | BC146767.1 | - | 4 - 5 | 457 | 9007211 | 64 |
D87072.1 | - | 4 - 5 | 457 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |