Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000593506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000593506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
ENST00000593506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021809.hg.1 |
ENST00000222374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021809.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021809.hg.1 |
TC19001592.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021810.hg.1 |
ENST00000222374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021810.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021810.hg.1 |
TC19001592.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021811.hg.1 |
ENST00000222374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021811.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021811.hg.1 |
TC19001592.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000593506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000593506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
ENST00000593506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010855.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900190081.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190081.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900190081.hg.1 |
TC1900010855.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900190081.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190081.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900190083.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190083.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900190083.hg.1 |
TC1900010855.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900190083.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190083.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900190080.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190080.hg.1 |
NM_145296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900190080.hg.1 |
TC1900010855.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900190080.hg.1 |
ENST00000222374.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900190080.hg.1 |
NM_145296.1 |
Refseq |