Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045437.hg.1 |
TC02002813.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045437.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045437.hg.1 |
ENST00000536361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045437.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045438.hg.1 |
TC02002813.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045438.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045438.hg.1 |
ENST00000536361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045438.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011229.hg.1 |
ENST00000435808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011229.hg.1 |
ENST00000435808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011229.hg.1 |
ENST00000435808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011229.hg.1 |
ENST00000435808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216540.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216540.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216540.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216540.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216540.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216540.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216540.hg.1 |
TC0200015838.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
ENST00000281828.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NM_005687.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015838.hg.1 |
NR_130154.1 |
Refseq |