Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023037.2 | NP_075463.2 | 28 - 29 | 4083 | 107 | |
XM_006719749.3 | XP_006719812.1 | 28 - 29 | 5164 | 107 | ||
XM_017020303.1 | XP_016875792.1 | 28 - 29 | 5641 | 107 | ||
XM_017020304.1 | XP_016875793.1 | 28 - 29 | 4083 | 107 | ||
XM_017020305.1 | XP_016875794.1 | 28 - 29 | 5660 | 107 | ||
XM_017020306.1 | XP_016875795.1 | 7 - 8 | 802 | 107 | ||
GenBank mRNA | AL049784.1 | - | 28 - 29 | 4083 | 107 | |
U50534.1 | - | 28 - 29 | 4082 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |