Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033180.hg.1 |
TC03001833.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033180.hg.1 |
ENST00000232217.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033180.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033181.hg.1 |
TC03001833.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033181.hg.1 |
ENST00000232217.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033181.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000506825.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000506825.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000506825.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033179.hg.1 |
TC03001833.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033179.hg.1 |
ENST00000232217.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033179.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194229.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194229.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194229.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194229.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194229.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194229.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194229.hg.1 |
TC0300012597.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300194231.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194231.hg.1 |
ENST00000506825.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194231.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194231.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194231.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194231.hg.1 |
ENST00000506825.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194231.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194231.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194231.hg.1 |
TC0300012597.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000506825.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000506825.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000506825.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012597.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194230.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194230.hg.1 |
ENST00000506825.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194230.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194230.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194230.hg.1 |
ENST00000232217.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194230.hg.1 |
ENST00000506825.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194230.hg.1 |
ENST00000511956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194230.hg.1 |
NM_004164.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194230.hg.1 |
TC0300012597.hg.1 |
Clariom D TC |