Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000354547.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000356340.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000357811.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000398964.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000398966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000398968.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000398969.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000398971.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000398972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000398973.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000398974.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000398978.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000517713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000520133.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000520267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
ENST00000522165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
NM_001130103.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
NM_080798.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
NM_080800.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
NM_080801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
NM_080802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
NM_080805.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004970.hg.1 |
TC10000424.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000354547.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000357811.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000398969.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000398975.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000398978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000456019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000479733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000517713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000520133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000520267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000522165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
NM_001130103.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
NM_080798.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
NM_080800.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
NM_080801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
NM_080802.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
NM_080805.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000354547.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000357811.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000398969.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000398975.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000398978.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000456019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000479733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000517713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000520133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000520267.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
ENST00000522165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
NM_001130103.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
NM_080798.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
NM_080800.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
NM_080801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
NM_080802.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
NM_080805.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153687.hg.1 |
TC1000007910.hg.1 |
Clariom D TC |