Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01112374_m1
See other GRAMD1B GE Assays ›
Gene Symbol:
GRAMD1B
Gene Name:
GRAM domain containing 1B
Gene Aliases:
-
Chromosome Location:
Chr.11: 123358299 - 123627771 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01112374_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: GRAMD1B
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_005271619.4
XP_005271676.1
16 - 17
2405
93
XM_005271621.4
XP_005271678.1
17 - 18
6792
93
XM_005271623.4
XP_005271680.1
16 - 17
2170
93
XM_005271624.4
XP_005271681.2
16 - 17
2085
93
XM_006718892.3
XP_006718955.1
17 - 18
2322
93
XM_011542926.2
XP_011541228.1
17 - 18
2319
93
XM_011542929.2
XP_011541231.1
16 - 17
2295
93
XM_011542930.2
XP_011541232.1
16 - 17
2936
93
XM_011542931.2
XP_011541233.1
16 - 17
2850
93
XM_017018034.1
XP_016873523.1
18 - 19
2390
93
XM_017018035.1
XP_016873524.1
18 - 19
2387
93
XM_017018036.1
XP_016873525.1
17 - 18
2857
93
XM_017018037.1
XP_016873526.1
18 - 19
3156
93
XM_017018039.1
XP_016873528.1
17 - 18
2448
93
XM_017018040.1
XP_016873529.1
16 - 17
2244
93
XM_017018042.1
XP_016873531.1
17 - 18
2448
93
XM_017018043.1
XP_016873532.1
16 - 17
2081
93
XM_017018044.1
XP_016873533.1
16 - 17
2405
93
XM_017018045.1
XP_016873534.1
17 - 18
6789
93
XM_017018046.1
XP_016873535.1
17 - 18
2894
93
XM_017018049.1
XP_016873538.1
17 - 18
2888
93
XM_017018050.1
XP_016873539.1
16 - 17
2164
93
GenBank mRNA
AK297606.1
-
16 - 17
2135
93
AY358924.1
-
6 - 7
1658
93
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Homo sapiens
Probe spans exons
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0