Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000512.hg.1 |
ENST00000270560.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000512.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000512.hg.1 |
TC17000050.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145450.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145450.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145450.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145450.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145450.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700145450.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700145450.hg.1 |
TC1700006642.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000270560.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
ENST00000576530.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006642.hg.1 |
NM_003963.2 |
Refseq |