Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000423645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000423645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000423645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000423645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000423645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000423645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002880.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002880.hg.1 |
ENST00000375378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002880.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002880.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002880.hg.1 |
TC20000229.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000149018.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149018.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149018.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149018.hg.1 |
TC2000007150.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149018.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149018.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149018.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000253354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000423645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000423645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000423645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000423645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000423645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000423645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
ENST00000464032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007150.hg.1 |
NM_033197.2 |
Refseq |