Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003968.hg.1 |
TC01000253.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003968.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003968.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000466697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000466697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000466697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000466697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000481686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
ENST00000481686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000466697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000466697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000466697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000466697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000481686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
ENST00000481686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007184.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150029.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150029.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150029.hg.1 |
ENST00000375120.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150029.hg.1 |
NM_015207.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150029.hg.1 |
TC0100007184.hg.1 |
Clariom D TC |