Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025138.hg.1 |
TC01001626.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025138.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025138.hg.1 |
ENST00000445394.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025138.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000477868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000477868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000477868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000477868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000477868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000477868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000477868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000477868.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
ENST00000482484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011027.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180936.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180936.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180936.hg.1 |
ENST00000367435.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100180936.hg.1 |
NM_024529.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100180936.hg.1 |
TC0100011027.hg.1 |
Clariom D TC |