Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000560649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000560649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
ENST00000560649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010514.hg.1 |
ENST00000306730.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010514.hg.1 |
ENST00000558136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010514.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15010514.hg.1 |
TC15001189.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500164717.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164717.hg.1 |
ENST00000558136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164717.hg.1 |
ENST00000560649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164717.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500164717.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164717.hg.1 |
ENST00000558136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164717.hg.1 |
ENST00000560649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500164717.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500164717.hg.1 |
TC1500008971.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000306730.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000558136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000560649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000560649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
ENST00000560649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500008971.hg.1 |
NM_020371.2 |
Refseq |