Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEBPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005760.2 | NP_005751.2 | 9 - 10 | 2475 | 97 | |
GenBank mRNA | AK312322.1 | - | 9 - 10 | 2474 | 97 | |
BC034475.2 | - | 9 - 10 | 2479 | 4822708 | 97 | |
M37197.1 | - | 9 - 10 | 2456 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |