Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057051.hg.1 |
TC01003717.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057051.hg.1 |
ENST00000255416.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057051.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016982.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228656.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228656.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228656.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228656.hg.1 |
ENST00000255416.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228656.hg.1 |
ENST00000621380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228656.hg.1 |
NM_004997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228656.hg.1 |
TC0100016982.hg.1 |
Clariom D TC |