Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014139.hg.1 |
ENST00000396398.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014139.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014139.hg.1 |
ENST00000403363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014139.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014139.hg.1 |
TC22000838.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014141.hg.1 |
ENST00000396398.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014141.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014141.hg.1 |
ENST00000403363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014141.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014141.hg.1 |
TC22000838.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014140.hg.1 |
ENST00000396398.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014140.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014140.hg.1 |
ENST00000403363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014140.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014140.hg.1 |
TC22000838.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200164148.hg.1 |
TC2200008856.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200164148.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164148.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164148.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164148.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200164148.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164148.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164148.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164148.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008856.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200164149.hg.1 |
TC2200008856.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200164149.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164149.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164149.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164149.hg.1 |
NM_000262.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200164149.hg.1 |
ENST00000396398.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164149.hg.1 |
ENST00000402937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164149.hg.1 |
ENST00000403363.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164149.hg.1 |
NM_000262.2 |
Refseq |