Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290263.1 | NP_001277192.1 | 25 - 26 | 2951 | 80 | |
NM_014689.2 | NP_055504.2 | 25 - 26 | 3094 | 80 | ||
XM_005246673.4 | XP_005246730.1 | 25 - 26 | 2953 | 80 | ||
XM_006712617.3 | XP_006712680.1 | 25 - 26 | 3118 | 80 | ||
XM_017004441.1 | XP_016859930.1 | 25 - 26 | 3121 | 80 | ||
XM_017004442.1 | XP_016859931.1 | 25 - 26 | 3124 | 80 | ||
XM_017004443.1 | XP_016859932.1 | 25 - 26 | 3170 | 80 | ||
XM_017004444.1 | XP_016859933.1 | 25 - 26 | 3122 | 80 | ||
XM_017004445.1 | XP_016859934.1 | 25 - 26 | 2951 | 80 | ||
XM_017004446.1 | XP_016859935.1 | 25 - 26 | 3122 | 80 | ||
XM_017004447.1 | XP_016859936.1 | 25 - 26 | 3120 | 80 | ||
GenBank mRNA | EU236710.2 | - | 25 - 26 | 2948 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |