Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258340.1 | NP_001245269.1 | 11 - 12 | 1503 | 89 | |
NM_001258341.1 | NP_001245270.1 | 11 - 12 | 1407 | 89 | ||
XM_017000262.1 | XP_016855751.1 | 10 - 11 | 1380 | 89 | ||
XM_017000263.1 | XP_016855752.1 | 11 - 12 | 1440 | 89 | ||
XM_017000264.1 | XP_016855753.1 | 11 - 12 | 1494 | 89 | ||
GenBank mRNA | AK313574.1 | - | 11 - 12 | 1391 | 89 | |
AK316123.1 | - | 13 - 14 | 1348 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Homo sapiens Probe spans exons |