Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056414.hg.1 |
TC01003683.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056414.hg.1 |
ENST00000332129.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056414.hg.1 |
ENST00000360529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056414.hg.1 |
ENST00000422435.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056414.hg.1 |
ENST00000461742.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056414.hg.1 |
ENST00000539858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056414.hg.1 |
NM_001252100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056414.hg.1 |
NM_001252102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056414.hg.1 |
NM_001252103.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01056414.hg.1 |
NM_017596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
ENST00000332129.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
ENST00000360529.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
ENST00000422435.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
ENST00000461742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
NM_001252100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
NM_001252102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
NM_001252103.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
NM_017596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
ENST00000332129.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
ENST00000360529.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
ENST00000422435.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
ENST00000461742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
NM_001252100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
NM_001252102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
NM_001252103.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
NM_017596.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100227800.hg.1 |
TC0100016899.hg.1 |
Clariom D TC |