Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000308528.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000308528.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000308528.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000308528.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000308528.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000308528.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000308528.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000308528.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000308528.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000308528.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000409448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000409448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000409448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000409448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000409448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000409448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000409448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000409448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000409448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
ENST00000409448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
NM_174898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
NM_174898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
NM_174898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
NM_174898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
NM_174898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
NM_174898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
NM_174898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
NM_174898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
NM_174898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013606.hg.1 |
NM_174898.2 |
Refseq |