Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002243.hg.1 |
TC05000190.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002243.hg.1 |
ENST00000281623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002243.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000504463.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000504463.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000504463.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000504463.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000505566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000505566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000505566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000505566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000506496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000506496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000506496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000513496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000513496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000513496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000513496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
ENST00000513496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500149008.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500149008.hg.1 |
ENST00000505566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500149008.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500149008.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500149008.hg.1 |
ENST00000505566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500149008.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500149008.hg.1 |
TC0500007251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000281623.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000296812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000504463.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000504463.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000504463.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000504463.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000505566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000505566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000505566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000505566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000506496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000506496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000506496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000509134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000513496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000513496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000513496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000513496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
ENST00000513496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_001297437.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_012176.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007251.hg.1 |
NM_033484.2 |
Refseq |