Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127399.1 | NP_001120871.1 | 2 - 3 | 232 | 129 | |
GenBank mRNA | AK025730.1 | - | 2 - 3 | 109 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |