Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012573.hg.1 |
TC09000940.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012573.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012573.hg.1 |
ENST00000341998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012573.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012573.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166154.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166154.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166154.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166154.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166154.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166154.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166154.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166154.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166154.hg.1 |
TC0900009660.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000286344.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
ENST00000341998.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_001193288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009660.hg.1 |
NM_020344.3 |
Refseq |