Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029373.hg.1 |
TC06002343.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029373.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029373.hg.1 |
ENST00000392093.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029373.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029373.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029372.hg.1 |
TC06002343.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029372.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029372.hg.1 |
ENST00000392093.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029372.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029372.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600196775.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196775.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196775.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600196775.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196775.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196775.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600196775.hg.1 |
TC0600014044.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600196776.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196776.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196776.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600196776.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196776.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600196776.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600196776.hg.1 |
TC0600014044.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000262193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
ENST00000462957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014044.hg.1 |
NM_002793.3 |
Refseq |