Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000327284.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000370010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000424947.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000436928.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000447266.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000483859.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000485497.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
NM_001100409.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
NM_001304792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
NM_015571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000327284.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000370010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000424947.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000436928.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000447266.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000483859.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
ENST00000485497.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
NM_001100409.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
NM_001304792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
NM_015571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159540.hg.1 |
TC0600008562.hg.1 |
Clariom D TC |