Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002856.hg.1 |
ENST00000397820.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002856.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002856.hg.1 |
TC19000187.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900150012.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150012.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150012.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150012.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150012.hg.1 |
TC1900007001.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900150012.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150012.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150012.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150012.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000397820.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000587494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
ENST00000592854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007001.hg.1 |
NM_001136482.2 |
Refseq |