Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000514301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000514301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000514301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000514301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
ENST00000514301.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156744.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156744.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156744.hg.1 |
ENST00000514301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156744.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156744.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156744.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156744.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156744.hg.1 |
ENST00000514301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156744.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156744.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156744.hg.1 |
TC0400008240.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000296414.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000507994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000512369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000514301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000514301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000514301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000514301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
ENST00000514301.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_001306151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008240.hg.1 |
NM_014395.2 |
Refseq |