Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011713.hg.1 |
TC0X002326.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011713.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011713.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000488351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000488351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000488351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000488351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
ENST00000488351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178580.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178580.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178580.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178580.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178580.hg.1 |
TC0X00011354.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000297904.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000488351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000488351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000488351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000488351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
ENST00000488351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011354.hg.1 |
NM_004469.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178613.hg.1 |
NR_037859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178613.hg.1 |
NR_037859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178613.hg.1 |
TC0X00011356.hg.1 |
Clariom D TC |