Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150500.hg.1 |
TC2200007186.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150500.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150500.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150500.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150500.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150500.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150500.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150499.hg.1 |
TC2200007186.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150499.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150499.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150499.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150499.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150499.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150499.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150501.hg.1 |
TC2200007186.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150501.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150501.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150501.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150501.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150501.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150501.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150495.hg.1 |
TC2200007186.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150495.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150495.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150495.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150495.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150495.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150495.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150497.hg.1 |
TC2200007186.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150497.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150497.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150497.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150497.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150497.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150497.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000308700.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
ENST00000404699.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007186.hg.1 |
NM_001303508.1 |
Refseq |