Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011947.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021924.hg.1 |
TC09001718.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021924.hg.1 |
ENST00000371763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021924.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021925.hg.1 |
TC09001718.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021925.hg.1 |
ENST00000371763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021925.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021926.hg.1 |
TC09001718.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021926.hg.1 |
ENST00000371763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021926.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181642.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181642.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181642.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181642.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181642.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181642.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181642.hg.1 |
TC0900011947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900181643.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181643.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181643.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181643.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181643.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181643.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181643.hg.1 |
TC0900011947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900181645.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181645.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181645.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181645.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181645.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181645.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181645.hg.1 |
TC0900011947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
ENST00000371763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
ENST00000613244.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
NM_182974.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011947.hg.1 |
NM_182974.2 |
Refseq |