Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008178.hg.1 |
TC05000580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008178.hg.1 |
ENST00000407847.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008178.hg.1 |
ENST00000464424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008178.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008174.hg.1 |
TC05000580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008174.hg.1 |
ENST00000407847.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008174.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008176.hg.1 |
TC05000580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008176.hg.1 |
ENST00000407847.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008176.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011877.hg.1 |
ENST00000442777.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011877.hg.1 |
ENST00000442777.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011877.hg.1 |
ENST00000442777.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011877.hg.1 |
ENST00000458103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011877.hg.1 |
ENST00000458103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008175.hg.1 |
TC05000580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008175.hg.1 |
ENST00000407847.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05008175.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000427739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000427739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000427739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000434521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000434521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000434521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000434521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000464424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000464424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
ENST00000464424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158297.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158297.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158297.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158297.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158297.hg.1 |
TC0500008491.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500158299.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158299.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158299.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158299.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158299.hg.1 |
TC0500008491.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011877.hg.1 |
ENST00000442777.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011877.hg.1 |
ENST00000442777.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011877.hg.1 |
ENST00000442777.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011877.hg.1 |
ENST00000458103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011877.hg.1 |
ENST00000458103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000427739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000427739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000427739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000434521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000434521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000434521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000434521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000464424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000464424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
ENST00000464424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008491.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158302.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158302.hg.1 |
ENST00000434521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158302.hg.1 |
ENST00000464424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158302.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158302.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158302.hg.1 |
ENST00000434521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158302.hg.1 |
ENST00000464424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158302.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158302.hg.1 |
TC0500008491.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500158300.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158300.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158300.hg.1 |
ENST00000407847.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500158300.hg.1 |
NM_001136239.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500158300.hg.1 |
TC0500008491.hg.1 |
Clariom D TC |