Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL22A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152888.2 | NP_690848.1 | 52 - 53 | 4232 | 80 | |
XM_011516883.2 | XP_011515185.1 | 51 - 52 | 4250 | 80 | ||
XM_011516884.2 | XP_011515186.1 | 50 - 51 | 4211 | 80 | ||
XM_011516885.2 | XP_011515187.1 | 50 - 51 | 4196 | 80 | ||
XM_011516886.2 | XP_011515188.1 | 52 - 53 | 4162 | 80 | ||
XM_011516887.2 | XP_011515189.1 | 48 - 49 | 3130 | 80 | ||
XM_011516889.2 | XP_011515191.1 | 41 - 42 | 2371 | 80 | ||
XM_017013150.1 | XP_016868639.1 | 52 - 53 | 4123 | 80 | ||
XM_017013151.1 | XP_016868640.1 | 48 - 49 | 4088 | 80 | ||
XM_017013152.1 | XP_016868641.1 | 47 - 48 | 3120 | 80 | ||
GenBank mRNA | AF406780.1 | - | 52 - 53 | 4219 | 80 | |
BC042075.1 | - | 26 - 27 | 1457 | 80 | ||
BC144535.1 | - | 40 - 41 | 2890 | 9053062 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |