Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000360379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000360379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000360379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000360379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000360379.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368756.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
ENST00000368758.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
NM_006945.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
NM_006945.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
NM_006945.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
NM_006945.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015847.hg.1 |
NM_006945.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000360379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000360379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000360379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000360379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000360379.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368756.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368757.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
ENST00000368758.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
NM_006945.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
NM_006945.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
NM_006945.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
NM_006945.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015847.hg.1 |
NM_006945.4 |
Refseq |