Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007733.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007733.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007733.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007733.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007733.hg.1 |
NR_046514.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007733.hg.1 |
NR_046514.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007733.hg.1 |
NR_046514.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007733.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007733.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007733.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007733.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007733.hg.1 |
NR_046514.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007733.hg.1 |
NR_046514.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007733.hg.1 |
NR_046514.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152153.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152153.hg.1 |
NR_046514.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152153.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152153.hg.1 |
NR_046514.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152153.hg.1 |
TC1300007733.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300152154.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152154.hg.1 |
NR_046514.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152154.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152154.hg.1 |
NR_046514.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152154.hg.1 |
TC1300007733.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300152155.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152155.hg.1 |
NR_046514.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152155.hg.1 |
ENST00000438290.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152155.hg.1 |
NR_046514.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152155.hg.1 |
TC1300007733.hg.1 |
Clariom D TC |