Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007825.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155673.hg.1 |
TC2200007825.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200155673.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155673.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155673.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155673.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155673.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155673.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155673.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155673.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155679.hg.1 |
TC2200007825.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200155679.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155679.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155679.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155679.hg.1 |
NM_033517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155679.hg.1 |
ENST00000262795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155679.hg.1 |
ENST00000414786.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155679.hg.1 |
ENST00000445220.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155679.hg.1 |
NM_033517.1 |
Refseq |