Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022496.hg.1 |
TC03001306.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022496.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022496.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022495.hg.1 |
TC03001306.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022495.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022495.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022498.hg.1 |
TC03001306.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022498.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022498.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178667.hg.1 |
TC0300010790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178666.hg.1 |
TC0300010790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300178671.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178671.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178671.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178671.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178671.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178671.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178671.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178671.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178671.hg.1 |
TC0300010790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000439293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
ENST00000452768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010790.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178670.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178670.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178670.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178670.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178670.hg.1 |
ENST00000420850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178670.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178670.hg.1 |
ENST00000425156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178670.hg.1 |
NR_040100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178670.hg.1 |
TC0300010790.hg.1 |
Clariom D TC |