Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001662.hg.1 |
ENST00000318578.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001662.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001662.hg.1 |
TC15000268.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001665.hg.1 |
ENST00000318578.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001665.hg.1 |
ENST00000561223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001665.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001665.hg.1 |
TC15000268.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001668.hg.1 |
ENST00000318578.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001668.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001668.hg.1 |
TC15000268.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
ENST00000561223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
ENST00000561223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006920.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001667.hg.1 |
ENST00000318578.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001667.hg.1 |
ENST00000561223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001667.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001667.hg.1 |
TC15000268.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001666.hg.1 |
ENST00000318578.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001666.hg.1 |
ENST00000561223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001666.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001666.hg.1 |
TC15000268.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001664.hg.1 |
ENST00000318578.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001664.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001664.hg.1 |
TC15000268.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001663.hg.1 |
ENST00000318578.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001663.hg.1 |
ENST00000561223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001663.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001663.hg.1 |
TC15000268.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
ENST00000561223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
ENST00000561223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006920.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147884.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147884.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147884.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147884.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147884.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147884.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147884.hg.1 |
TC1500006920.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500147887.hg.1 |
ENST00000561223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147887.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147887.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147887.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147887.hg.1 |
ENST00000561223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147887.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147887.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147887.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147887.hg.1 |
TC1500006920.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500147886.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147886.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147886.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147886.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147886.hg.1 |
TC1500006920.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500147885.hg.1 |
ENST00000561223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147885.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147885.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147885.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147885.hg.1 |
ENST00000561223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147885.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147885.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147885.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147885.hg.1 |
TC1500006920.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500147888.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147888.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147888.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147888.hg.1 |
ENST00000625107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147888.hg.1 |
NM_001302797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147888.hg.1 |
NM_207445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147888.hg.1 |
TC1500006920.hg.1 |
Clariom D TC |