Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167962.1 | NP_001161434.1 | 14 - 15 | 1747 | 154 | |
NM_001321787.1 | NP_001308716.1 | 14 - 15 | 1747 | 154 | ||
NM_198999.2 | NP_945350.1 | 15 - 16 | 1843 | 154 | ||
NM_206883.2 | NP_996766.1 | 15 - 16 | 1843 | 154 | ||
NR_135801.1 | 13 - 14 | 1599 | 154 | |||
XM_011516170.2 | XP_011514472.1 | 15 - 16 | 2633 | 154 | ||
GenBank mRNA | AF523354.1 | - | 15 - 16 | 1817 | 154 | |
AY256823.1 | - | 15 - 16 | 1817 | 154 | ||
AY289133.1 | - | 13 - 14 | 1589 | 154 | ||
AY289134.1 | - | 12 - 13 | 1493 | 154 | ||
BC100833.1 | - | 13 - 14 | 1652 | 40001467 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons |