Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033814.hg.1 |
TC01002194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033814.hg.1 |
ENST00000400897.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033814.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400898.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400898.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400898.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000400898.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000478645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000478645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000478645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000480221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000480221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000480221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000480221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000480221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
ENST00000480221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_139208.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_139208.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_139208.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012868.hg.1 |
NM_139208.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400898.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400898.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400898.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000400898.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000478645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000478645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000478645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000480221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000480221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000480221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000480221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000480221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
ENST00000480221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_139208.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_139208.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_139208.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012868.hg.1 |
NM_139208.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194999.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194999.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194999.hg.1 |
ENST00000400897.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194999.hg.1 |
NM_006610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194999.hg.1 |
TC0100012868.hg.1 |
Clariom D TC |